Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163941.1 | NP_001157413.1 | 15 | 1989 | ||
NM_178559.5 | NP_848654.3 | 6 | 963 | |||
XM_011515367.2 | XP_011513669.1 | 7 | 909 | |||
GenBank mRNA | AB353947.1 | - | 15 | 1989 | ||
AF319622.1 | - | 1 | 29 | |||
AF399931.1 | - | 4 | 477 | |||
AY090613.1 | - | 6 | 884 | |||
AY230001.1 | - | 15 | 1882 | |||
AY234788.1 | - | 6 | 884 | |||
AY851365.1 | - | 6 | 770 | |||
BC104894.1 | - | 6 | 713 | 8143897 | ||
BC104920.1 | - | 6 | 711 | 8143923 | ||
GU437216.1 | - | 3 | 417 | |||
GU437217.1 | - | 3 | 417 |