Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178562.4 | NP_848657.1 | 7 | 897 | ||
XM_006715960.3 | XP_006716023.1 | 7 | 725 | |||
GenBank mRNA | AF276891.1 | - | 7 | 632 | ||
AK000208.1 | - | 2 | 511 | |||
AY236849.1 | - | 7 | 721 | |||
BC044244.1 | - | 7 | 861 | 5759132 |