Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081675.2 | NP_001075144.2 | 1, 2 | 1383 | ||
XM_005250901.4 | XP_005250958.1 | 2, 3 | 1604 | |||
GenBank mRNA | AB196636.1 | - | 1, 2 | 1359 | ||
BC127886.1 | - | 1, 2 | 1383 | 40131758 |