Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317942.1 | NP_001304871.1 | 2, 3 | 268 | ||
NM_178565.4 | NP_848660.3 | 3, 4 | 947 | |||
XM_011517018.1 | XP_011515320.1 | 3, 4 | 963 | |||
XM_011517019.1 | XP_011515321.1 | 3, 4 | 963 | |||
XM_017013395.1 | XP_016868884.1 | 3, 4 | 936 | |||
GenBank mRNA | AK123023.1 | - | 3, 4 | 910 | ||
AK123027.1 | - | 1, 2 | 77 | |||
AY358166.1 | - | 3, 4 | 600 | |||
BC027938.1 | - | 2, 3 | 268 | 5201681 |