Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010887.2 | NP_001010887.2 | 4, 5 | 662 | ||
XM_005251448.3 | XP_005251505.1 | 5, 6 | 713 | |||
XM_011517859.2 | XP_011516161.1 | 4, 5 | 555 | |||
XM_017014694.1 | XP_016870183.1 | 4, 5 | 418 | |||
GenBank mRNA | AK123581.1 | - | 4, 5 | 662 | ||
AY312516.1 | - | 4, 5 | 504 | |||
BC092487.1 | - | 5, 6 | 713 | 30528463 |