Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA2022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008537.2 | NP_001008537.1 | 3 | 3154 | ||
GenBank mRNA | AB095942.1 | - | 4 | 2782 | ||
AK128651.1 | - | 1 | 2146 | |||
AK307471.1 | - | 3 | 3148 | |||
AY563507.1 | - | 3 | 2851 | |||
BC152557.1 | - | 4 | 2782 | 8860127 |