Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002273.2 | NP_001002273.1 | 7 | 1417 | ||
NM_001002274.2 | NP_001002274.1 | 7 | 1420 | |||
NM_001002275.2 | NP_001002275.1 | 8 | 1474 | |||
NM_001190828.1 | NP_001177757.1 | 7 | 1456 | |||
NM_004001.4 | NP_003992.3 | 8 | 1477 | |||
XM_017000671.1 | XP_016856160.1 | 9 | 1441 | |||
XM_017000672.1 | XP_016856161.1 | 8 | 1384 | |||
GenBank mRNA | AB209585.1 | - | 6 | 4598 | ||
AK308150.1 | - | 7 | 1347 | |||
BC031992.1 | - | 8 | 1387 | 4752609 | ||
BC141826.1 | - | 7 | 1330 | 40148276 | ||
BC146678.1 | - | 7 | 1335 | 40148906 | ||
H04104.1 | - | 1 | 212 | |||
M28696.1 | - | 7 | 1333 |