Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127593.1 | NP_001121065.1 | 6 | 840 | ||
NM_001127595.1 | NP_001121067.1 | 6 | 822 | |||
NM_001127596.1 | NP_001121068.1 | 6 | 819 | |||
XM_011509293.2 | XP_011507595.2 | 4 | 471 | |||
GenBank mRNA | AK223268.1 | - | 5 | 701 | ||
AK223295.1 | - | 5 | 713 | |||
AK302126.1 | - | 5 | 1052 | |||
BC017865.1 | - | 5 | 733 | 4690249 | ||
BC033678.1 | - | 5 | 708 | 5180561 | ||
BC036723.1 | - | 5 | 977 | 5206097 | ||
KU178060.1 | - | 5 | 793 | |||
KU178061.1 | - | 4 | 535 | |||
M24853.1 | - | 3 | 595 | |||
S62490.1 | - | 1 | 85 | |||
S76824.1 | - | 3 | 622 | |||
X52645.1 | - | 5 | 718 |
Gene Symbol: FCGR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000570.4 | NP_000561.3 | 6 | 807 | ||
NM_001244753.1 | NP_001231682.1 | 5 | 1053 | |||
NM_001271035.1 | NP_001257964.1 | 5 | 1050 | |||
NM_001271036.1 | NP_001257965.1 | 5 | 919 | |||
NM_001271037.1 | NP_001257966.1 | 4 | 661 | |||
GenBank mRNA | AB025256.1 | - | 5 | 718 | ||
AJ581669.1 | - | 5 | 718 | |||
AK313219.1 | - | 5 | 799 | |||
BC128562.1 | - | 5 | 702 | 40126686 | ||
J04162.1 | - | 5 | 702 | |||
M24854.1 | - | 5 | 783 | |||
X07934.1 | - | 5 | 718 | |||
X16863.1 | - | 5 | 718 |