Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272046.1 | NP_001258975.1 | 11 | 1844 | ||
NM_001320804.1 | NP_001307733.1 | 11 | 1786 | |||
XM_011539754.2 | XP_011538056.1 | 12 | 2021 | |||
XM_011539757.2 | XP_011538059.1 | 11 | 2116 | |||
XM_017016177.1 | XP_016871666.1 | 11 | 1596 | |||
XM_017016178.1 | XP_016871667.1 | 11 | 1596 | |||
XM_017016179.1 | XP_016871668.1 | 11 | 1549 | |||
XM_017016180.1 | XP_016871669.1 | 3 | 692 | |||
GenBank mRNA | AJ536328.2 | - | 11 | 1768 | ||
AK122716.1 | - | 11 | 1839 | |||
AK127756.1 | - | 11 | 1675 | |||
AY572972.1 | - | 11 | 1606 | |||
BC128588.1 | - | 10 | 1612 | 40128488 | ||
BC143380.1 | - | 11 | 1546 |