Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272046.1 | NP_001258975.1 | 3 | 428 | ||
NM_001320804.1 | NP_001307733.1 | 3 | 370 | |||
XM_011539754.2 | XP_011538056.1 | 3 | 427 | |||
XM_011539757.2 | XP_011538059.1 | 3 | 700 | |||
XM_017016177.1 | XP_016871666.1 | 3 | 180 | |||
XM_017016178.1 | XP_016871667.1 | 3 | 189 | |||
XM_017016179.1 | XP_016871668.1 | 3 | 133 | |||
XM_017016181.1 | XP_016871670.1 | 3 | 177 | |||
GenBank mRNA | AJ536328.2 | - | 3 | 352 | ||
AJ616914.1 | - | 3 | 352 | |||
AK122716.1 | - | 3 | 423 | |||
AK127756.1 | - | 3 | 259 | |||
AY572972.1 | - | 3 | 190 | |||
BC128588.1 | - | 3 | 326 | 40128488 | ||
BC143380.1 | - | 3 | 130 | |||
DA170480.1 | - | 3 | 428 | |||
DB210781.1 | - | 3 | 428 |