Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000570.4 | NP_000561.3 | 4 | 340 | ||
NM_001244753.1 | NP_001231682.1 | 3 | 586 | |||
NM_001271035.1 | NP_001257964.1 | 3 | 583 | |||
NM_001271036.1 | NP_001257965.1 | 3 | 452 | |||
NM_001271037.1 | NP_001257966.1 | 3 | 452 | |||
GenBank mRNA | AB025256.1 | - | 3 | 251 | ||
AJ581669.1 | - | 3 | 251 | |||
AK313219.1 | - | 3 | 332 | |||
AK316565.1 | - | 3 | 272 | |||
BC128562.1 | - | 3 | 235 | 40126686 | ||
DA670720.1 | - | 3 | 452 | |||
DA671001.1 | - | 3 | 437 | |||
DA672751.1 | - | 4 | 340 | |||
DA672763.1 | - | 3 | 492 | |||
DA673839.1 | - | 3 | 349 | |||
J04162.1 | - | 3 | 235 | |||
M24854.1 | - | 3 | 316 | |||
X07934.1 | - | 3 | 251 | |||
X16863.1 | - | 3 | 251 |
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127593.1 | NP_001121065.1 | 4 | 373 | ||
NM_001127595.1 | NP_001121067.1 | 4 | 355 | |||
NM_001127596.1 | NP_001121068.1 | 4 | 352 | |||
XM_011509293.2 | XP_011507595.2 | 3 | 262 | |||
GenBank mRNA | AK223268.1 | - | 3 | 234 | ||
AK223295.1 | - | 3 | 246 | |||
AK302126.1 | - | 3 | 585 | |||
BC017865.1 | - | 3 | 266 | 4690249 | ||
BC033678.1 | - | 3 | 241 | 5180561 | ||
BC036723.1 | - | 3 | 510 | 5206097 | ||
BI768140.1 | - | 3 | 510 | |||
DA674960.1 | - | 4 | 332 | |||
DA933259.1 | - | 4 | 346 | |||
DA944383.1 | - | 3 | 279 | |||
DA957430.1 | - | 4 | 371 | |||
KU178060.1 | - | 3 | 326 | |||
KU178061.1 | - | 3 | 326 | |||
M24853.1 | - | 1 | 128 | |||
S76824.1 | - | 1 | 155 | |||
X52645.1 | - | 3 | 251 |