Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136019.2 | NP_001129491.1 | 7 | 1775 | ||
NM_004107.4 | NP_004098.1 | 6 | 1511 | |||
XM_005258657.1 | XP_005258714.1 | 6 | 1330 | |||
XM_011526627.2 | XP_011524929.1 | 5 | 1192 | |||
GenBank mRNA | AK074734.1 | - | 3 | 2653 | ||
AK075510.1 | - | 7 | 1709 | |||
BC008734.2 | - | 6 | 1455 | 3163446 | ||
BE245284.1 | - | 3 | 545 | |||
BX647163.1 | - | 7 | 1395 | |||
U12255.1 | - | 6 | 1414 |