Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1FNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181788.1 | NP_861453.1 | 1 | 580 | ||
GenBank mRNA | AB121028.2 | - | 1 | 220 | ||
AY302593.1 | - | 1 | 580 | |||
BC118635.1 | - | 1 | 528 | 40023772 | ||
BC119787.1 | - | 1 | 530 | 40023773 |