Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199289.2 | NP_954983.1 | 8, 9 | 701 | ||
XM_006719808.3 | XP_006719871.2 | 8, 9 | 782 | |||
XM_011535066.2 | XP_011533368.1 | 8, 9 | 782 | |||
XM_011535068.2 | XP_011533370.1 | 8, 9 | 782 | |||
XM_017020555.1 | XP_016876044.1 | 8, 9 | 784 | |||
XM_017020556.1 | XP_016876045.1 | 8, 9 | 788 | |||
XM_017020557.1 | XP_016876046.1 | 8, 9 | 792 | |||
GenBank mRNA | BC063885.1 | - | 8, 9 | 687 | 30383658 |