Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103184.3 | NP_001096654.1 | 4 | 2388 | ||
NM_001277313.1 | NP_001264242.1 | 8 | 3413 | |||
XM_011521504.2 | XP_011519806.1 | 8 | 3236 | |||
XM_011521505.2 | XP_011519807.1 | 8 | 3445 | |||
XM_011521506.2 | XP_011519808.1 | 6 | 3150 | |||
XM_011521507.2 | XP_011519809.1 | 8 | 3445 | |||
XM_011521509.2 | XP_011519811.1 | 7 | 1444 | |||
XM_011521511.2 | XP_011519813.1 | 5 | 2915 | |||
XM_017022130.1 | XP_016877619.1 | 5 | 3607 | |||
XM_017022131.1 | XP_016877620.1 | 8 | 3445 | |||
XM_017022132.1 | XP_016877621.1 | 5 | 3361 | |||
XM_017022133.1 | XP_016877622.1 | 7 | 3266 | |||
XM_017022134.1 | XP_016877623.1 | 5 | 1817 | |||
XM_017022135.1 | XP_016877624.1 | 5 | 3086 |