Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103184.3 | NP_001096654.1 | 10, 11 | 3126 | ||
NM_001277313.1 | NP_001264242.1 | 14, 15 | 4151 | |||
XM_011521504.2 | XP_011519806.1 | 14, 15 | 3974 | |||
XM_011521505.2 | XP_011519807.1 | 14, 15 | 4183 | |||
XM_011521506.2 | XP_011519808.1 | 12, 13 | 3888 | |||
XM_011521507.2 | XP_011519809.1 | 14, 15 | 4183 | |||
XM_011521509.2 | XP_011519811.1 | 13, 14 | 2182 | |||
XM_011521511.2 | XP_011519813.1 | 11, 12 | 3653 | |||
XM_017022130.1 | XP_016877619.1 | 11, 12 | 4345 | |||
XM_017022131.1 | XP_016877620.1 | 14, 15 | 4183 | |||
XM_017022132.1 | XP_016877621.1 | 11, 12 | 4099 | |||
XM_017022133.1 | XP_016877622.1 | 13, 14 | 4004 | |||
XM_017022134.1 | XP_016877623.1 | 11, 12 | 2555 | |||
XM_017022135.1 | XP_016877624.1 | 11, 12 | 3824 |