Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181449.2 | NP_852114.2 | 2 | 264 | ||
XM_017024575.1 | XP_016880064.1 | 2 | 464 | |||
GenBank mRNA | AF395839.1 | - | 2 | 168 | ||
AK298031.1 | - | 2 | 163 | |||
AK303545.1 | - | 2 | 264 | |||
BC100888.1 | - | 2 | 162 | 40003374 | ||
BC100889.1 | - | 2 | 162 | 40003375 | ||
BC100890.1 | - | 2 | 162 | 40003376 | ||
BX648376.1 | - | 2 | 246 |