Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321116.1 | NP_001308045.1 | 3 | 732 | ||
NM_001321117.1 | NP_001308046.1 | 3 | 721 | |||
NM_181846.2 | NP_862829.1 | 3 | 737 | |||
XM_006723192.3 | XP_006723255.1 | 3 | 709 | |||
XM_011526917.2 | XP_011525219.1 | 3 | 1815 | |||
XM_011526918.2 | XP_011525220.1 | 2 | 255 | |||
XM_017026746.1 | XP_016882235.1 | 3 | 735 | |||
XM_017026747.1 | XP_016882236.1 | 3 | 688 | |||
GenBank mRNA | AK128716.1 | - | 3 | 727 | ||
BC101630.1 | - | 2 | 650 | 8069136 | ||
BC112277.1 | - | 2 | 649 | 8327745 | ||
BC143446.1 | - | 2 | 634 | |||
BX537741.1 | - | 3 | 737 |