Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010889.2 | NP_001010889.1 | 3 | 820 | ||
GenBank mRNA | BC101342.2 | - | 3 | 820 | 40025149 |
Gene Symbol: PRAMEF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146344.2 | NP_001139816.2 | 3 | 811 | ||
XM_011541479.2 | XP_011539781.1 | 2 | 997 | |||
GenBank mRNA | AL049680.1 | - | 3 | 810 | ||
BC144426.1 | - | 3 | 810 | 9052947 |
Gene Symbol: PRAMEF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098376.2 | NP_001091846.1 | 3 | 834 |
Gene Symbol: PRAMEF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310134.1 | NP_001297063.1 | 2 | 756 |
Gene Symbol: PRAMEF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306072.1 | NP_001293001.1 | 2 | 756 |
Gene Symbol: PRAMEF27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300891.1 | NP_001287820.1 | 2 | 740 |
Gene Symbol: PRAMEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009611.4 | NP_001009611.2 | 3 | 811 | ||
GenBank mRNA | AL049681.1 | - | 3 | 811 |
Gene Symbol: PRAMEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013407.2 | NP_001013425.2 | 3 | 834 | ||
GenBank mRNA | BC101344.1 | - | 3 | 820 | 40025152 | |
BC130274.1 | - | 3 | 819 | 40146305 | ||
BC130276.1 | - | 3 | 819 | 40146307 |
Gene Symbol: PRAMEF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010890.2 | NP_001010890.2 | 2 | 740 | ||
GenBank mRNA | AK124292.1 | - | 3 | 834 | ||
BC101343.2 | - | 3 | 820 | 40025151 | ||
BC137391.1 | - | 3 | 820 | 9021389 | ||
BC171794.1 | - | 3 | 820 | 9054448 |