Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136561.3 | NP_001130033.3 | 2 | 592 | ||
GenBank mRNA | BC144551.1 | - | 2 | 592 | 9053078 | |
BC157858.1 | - | 1 | 444 | 9057077 | ||
BC171863.1 | - | 1 | 444 | 9054517 |
Gene Symbol: HNRNPCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013631.2 | NP_001013653.1 | 2 | 606 | ||
GenBank mRNA | BC002696.2 | - | 2 | 575 | ||
BC137258.1 | - | 2 | 606 | 9021255 | ||
BC171866.1 | - | 1 | 444 | 9054520 |