Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRAMEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009611.4 | NP_001009611.2 | 4 | 1402 | ||
GenBank mRNA | AL049681.1 | - | 4 | 1402 |
Gene Symbol: PRAMEF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC144426.1 | - | 4 | 1401 | 9052947 |
Gene Symbol: PRAMEF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098376.2 | NP_001091846.1 | 4 | 1425 |
Gene Symbol: PRAMEF27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300891.1 | NP_001287820.1 | 3 | 1331 |
Gene Symbol: PRAMEF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010890.2 | NP_001010890.2 | 3 | 1331 | ||
GenBank mRNA | AK124292.1 | - | 4 | 1425 | ||
BC101343.2 | - | 4 | 1411 | 40025151 | ||
BC137391.1 | - | 4 | 1411 | 9021389 | ||
BC171794.1 | - | 4 | 1411 | 9054448 |