Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2T8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005522.1 | NP_001005522.1 | 1 | 772 | ||
XM_011544178.2 | XP_011542480.1 | 2 | 2115 | |||
XM_017001172.1 | XP_016856661.1 | 2 | 4894 |
Gene Symbol: OR2T12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004692.1 | NP_001004692.1 | 1 | 772 |
Gene Symbol: OR2T33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004695.1 | NP_001004695.1 | 1 | 772 | ||
GenBank mRNA | BC136994.1 | - | 1 | 794 | 9020986 | |
BC136995.1 | - | 1 | 794 | 9020987 |