Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004310.2 | NP_001004310.2 | 6 | 1023 | ||
NM_001284217.1 | NP_001271146.1 | 7 | 1089 | |||
XM_005245128.3 | XP_005245185.1 | 7 | 1081 | |||
XM_005245129.3 | XP_005245186.1 | 6 | 1048 | |||
XM_005245131.3 | XP_005245188.1 | 7 | 1099 | |||
XM_006711292.3 | XP_006711355.1 | 7 | 1099 | |||
XM_011509480.1 | XP_011507782.1 | 7 | 1088 | |||
XM_011509481.2 | XP_011507783.1 | 7 | 1099 | |||
XM_017001176.1 | XP_016856665.1 | 7 | 952 | |||
XM_017001177.1 | XP_016856666.1 | 6 | 885 | |||
GenBank mRNA | AK131201.1 | - | 7 | 1088 | ||
AY212514.1 | - | 6 | 989 | |||
AY513661.1 | - | 6 | 981 | |||
AY654627.1 | - | 5 | 697 | |||
AY654628.1 | - | 6 | 982 |