Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004310.2 | NP_001004310.2 | 9, 10 | 1229 | ||
XM_005245128.3 | XP_005245185.1 | 10, 11 | 1287 | |||
XM_005245129.3 | XP_005245186.1 | 9, 10 | 1254 | |||
XM_005245131.3 | XP_005245188.1 | 9, 10 | 1273 | |||
XM_011509480.1 | XP_011507782.1 | 10, 11 | 1294 | |||
XM_017001176.1 | XP_016856665.1 | 10, 11 | 1158 | |||
XM_017001177.1 | XP_016856666.1 | 9, 10 | 1091 | |||
GenBank mRNA | AY212514.1 | - | 8, 9 | 1163 | ||
AY513661.1 | - | 9, 10 | 1187 | |||
AY654628.1 | - | 9, 10 | 1188 |