Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258012.3 | NP_001244941.2 | 7 | 872 | ||
NM_001258013.3 | NP_001244942.2 | 8 | 836 | |||
NM_001258014.3 | NP_001244943.2 | 7 | 719 | |||
NM_001258015.2 | NP_001244944.1 | 6 | 623 | |||
NM_001258016.3 | NP_001244945.2 | 6 | 688 | |||
NM_004110.5 | NP_004101.3 | 7 | 761 | |||
NM_024417.4 | NP_077728.3 | 7 | 743 | |||
NR_047576.2 | 7 | 843 | ||||
XM_006721772.1 | XP_006721835.1 | 7 | 691 | |||
XM_011524527.1 | XP_011522829.1 | 7 | 709 | |||
XM_011524528.1 | XP_011522830.1 | 7 | 798 | |||
XM_011524529.1 | XP_011522831.1 | 7 | 801 | |||
XM_011524531.2 | XP_011522833.1 | 6 | 694 | |||
XM_011524532.2 | XP_011522834.1 | 6 | 701 | |||
XM_011524533.2 | XP_011522835.1 | 6 | 669 | |||
GenBank mRNA | AK094120.1 | - | 8 | 941 | ||
AK098648.1 | - | 7 | 680 | |||
AK223530.1 | - | 7 | 673 | |||
AK293208.1 | - | 8 | 781 | |||
AK293210.1 | - | 7 | 780 | |||
AK295307.1 | - | 7 | 872 | |||
AK298908.1 | - | 6 | 566 | |||
AK301779.1 | - | 7 | 656 | |||
AK301977.1 | - | 6 | 688 | |||
BC002960.1 | - | 7 | 682 | 3537008 | ||
BC018911.2 | - | 7 | 689 | |||
BC034763.1 | - | 7 | 1344 | |||
BC063493.1 | - | 7 | 681 | 5404814 | ||
BT006684.1 | - | 7 | 656 | |||
J03826.1 | - | 7 | 694 |