Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007178.3 | NP_009109.3 | 6 | 869 | ||
XM_005253294.1 | XP_005253351.1 | 5 | 731 | |||
GenBank mRNA | AB024327.1 | - | 6 | 815 | ||
AF161496.1 | - | 6 | 816 | |||
AJ010025.1 | - | 6 | 662 | |||
AK297942.1 | - | 7 | 868 | |||
AK312295.1 | - | 6 | 810 | |||
AK315891.1 | - | 5 | 696 | |||
AL136691.1 | - | 6 | 811 | |||
AY049776.1 | - | 6 | 562 | |||
BC000162.2 | - | 6 | 660 | 2900954 | ||
BC062306.1 | - | 6 | 824 | 3349549 | ||
BT020044.1 | - | 6 | 515 | |||
BT020045.1 | - | 6 | 515 |