Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038705.1 | NP_001033794.1 | 3 | 1575 | ||
GenBank mRNA | AY255534.1 | - | 3 | 1010 | ||
DQ318360.1 | - | 3 | 1575 |
Set Membership: |
G-protein coupled receptor |