Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308028.1 | NP_001294957.1 | 8 | 1223 | ||
NM_005246.3 | NP_005237.2 | 10 | 1609 | |||
XM_011543266.1 | XP_011541568.1 | 10 | 1462 | |||
XM_011543267.1 | XP_011541569.1 | 10 | 3346 | |||
XM_011543269.1 | XP_011541571.1 | 9 | 3200 | |||
XM_011543270.2 | XP_011541572.1 | 9 | 1377 | |||
XM_011543271.2 | XP_011541573.1 | 10 | 1462 | |||
XM_011543272.1 | XP_011541574.1 | 10 | 1462 | |||
XM_011543278.1 | XP_011541580.1 | 10 | 1462 | |||
XM_017009229.1 | XP_016864718.1 | 11 | 1609 | |||
XM_017009230.1 | XP_016864719.1 | 9 | 1377 | |||
XM_017009231.1 | XP_016864720.1 | 9 | 3200 | |||
XM_017009232.1 | XP_016864721.1 | 9 | 1250 | |||
XM_017009233.1 | XP_016864722.1 | 8 | 1193 | |||
XM_017009234.1 | XP_016864723.1 | 10 | 1462 | |||
XM_017009235.1 | XP_016864724.1 | 10 | 1462 | |||
GenBank mRNA | AK299855.1 | - | 8 | 1188 | ||
AK315234.1 | - | 8 | 1297 | |||
BC017060.1 | - | 8 | 1291 | |||
J03358.1 | - | 10 | 1609 |