Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042575.2 | NP_001036040.2 | 16 | 2256 | ||
NR_026734.1 | 17 | 2369 | ||||
XM_011512754.1 | XP_011511056.1 | 17 | 2701 | |||
XM_011512758.1 | XP_011511060.1 | 16 | 2122 | |||
GenBank mRNA | AK131211.1 | - | 17 | 2369 | ||
BC117322.1 | - | 16 | 2168 | 40125873 |