Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198505.2 | NP_940907.2 | 27 | 3164 | ||
XM_017006304.1 | XP_016861793.1 | 23 | 2477 | |||
XM_017006305.1 | XP_016861794.1 | 14 | 1580 | |||
GenBank mRNA | AY823163.1 | - | 27 | 3164 |