Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198506.4 | NP_940908.3 | 4 | 1988 | ||
XM_017008167.1 | XP_016863656.1 | 3 | 1610 | |||
GenBank mRNA | AK126648.1 | - | 4 | 2185 | ||
BC104037.2 | - | 3 | 1649 | 40026462 |