Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRGM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145805.1 | NP_001139277.1 | 2 | 1337 | ||
XM_011537641.2 | XP_011535943.1 | 2 | 1337 | |||
GenBank mRNA | BC128167.1 | - | 1 | 217 | 40113046 | |
BC128168.1 | - | 1 | 224 | 40113047 | ||
EU742619.1 | - | 2 | 1337 |