Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010892.2 | NP_001010892.1 | 3 | 1745 | ||
NM_001161664.1 | NP_001155136.1 | 3 | 1745 | |||
XM_017010826.1 | XP_016866315.1 | 4 | 5312 | |||
GenBank mRNA | AK095293.1 | - | 4 | 1604 | ||
AK299754.1 | - | 3 | 1745 | |||
BC106949.2 | - | 3 | 1723 | 40034633 |