Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207366.2 | NP_997249.2 | 9 | 1219 | ||
XM_011515373.2 | XP_011513675.1 | 9 | 1203 | |||
GenBank mRNA | AK126048.1 | - | 4 | 507 | ||
AK301928.1 | - | 9 | 1182 | |||
DB087561.1 | - | 4 | 507 |