Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182489.1 | NP_872295.1 | 8 | 907 | ||
XM_011516137.2 | XP_011514439.1 | 8 | 1760 | |||
XM_011516138.2 | XP_011514440.1 | 8 | 1145 | |||
XM_011516139.2 | XP_011514441.1 | 9 | 1389 | |||
GenBank mRNA | AF513502.1 | - | 8 | 907 |