Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194313.2 | NP_919289.2 | 11 | 2549 | ||
XM_005251451.2 | XP_005251508.1 | 7 | 1442 | |||
XM_006716764.2 | XP_006716827.1 | 5 | 1153 | |||
XM_011517861.1 | XP_011516163.1 | 12 | 2770 | |||
XM_011517863.2 | XP_011516165.1 | 11 | 3859 | |||
XM_017014697.1 | XP_016870186.1 | 11 | 2863 | |||
XM_017014698.1 | XP_016870187.1 | 10 | 2636 | |||
GenBank mRNA | AK125872.1 | - | 2 | 925 | ||
BC110502.1 | - | 2 | 907 | |||
BC110503.1 | - | 2 | 907 | 40028587 |