Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MURC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018116.2 | NP_001018126.1 | 2 | 521 | ||
GenBank mRNA | AK128238.1 | - | 1 | 3721 | ||
AK300099.1 | - | 2 | 521 | |||
BC090888.1 | - | 2 | 438 | 30529833 | ||
EU487253.1 | - | 2 | 456 |