Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGAT2L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198512.2 | NP_940914.1 | 6 | 939 | ||
GenBank mRNA | AK129500.1 | - | 6 | 936 | ||
BG739793.1 | - | 1 | 112 |