Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177987.2 | NP_817124.1 | 4 | 621 | ||
XM_011519459.2 | XP_011517761.1 | 4 | 661 | |||
XM_011519460.2 | XP_011517762.1 | 3 | 307 | |||
XM_017016192.1 | XP_016871681.1 | 2 | 558 | |||
XM_017016193.1 | XP_016871682.1 | 2 | 378 | |||
GenBank mRNA | BC101270.2 | - | 4 | 614 | ||
BC101271.1 | - | 4 | 614 |