Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178019.2 | NP_821138.1 | 3 | 463 | ||
GenBank mRNA | AF432876.1 | - | 3 | 445 | ||
AK292427.1 | - | 3 | 462 | |||
BC101692.1 | - | 3 | 423 | 8069198 | ||
BC110384.1 | - | 3 | 463 | 5166215 | ||
BI827754.1 | - | 3 | 463 |