Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004464.3 | NP_004455.2 | 1 | 523 | ||
NM_033143.2 | NP_149134.1 | 1 | 523 | |||
GenBank mRNA | AA461028.1 | - | 1 | 518 | ||
AB016517.1 | - | 1 | 286 | |||
AF171928.1 | - | 1 | 436 | |||
AF355187.1 | - | 1 | 286 | |||
AF535149.1 | - | 1 | 448 | |||
AK291962.1 | - | 1 | 448 | |||
AK312065.1 | - | 1 | 472 | |||
BC074858.2 | - | 1 | 335 | 30915226 | ||
BC074859.2 | - | 1 | 335 | 30915463 | ||
BC131502.1 | - | 1 | 424 | 40082281 | ||
DR004226.1 | - | 1 | 573 | |||
M37825.1 | - | 1 | 425 |