Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NMNAT3 SIRNA Assays ›
Gene Symbol:
NMNAT3
Gene Name:
nicotinamide nucleotide adenylyltransferase 3
Gene Aliases:
FKSG76, PNAT-3, PNAT3
Chromosome Location:
Chr.3: 139560180 - 139678050 on Build GRCh38
UniGene ID:
Hs.208673
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51395
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NMNAT3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001200047.2
NP_001186976.1
4
866
NM_001320510.1
NP_001307439.1
7
1415
NM_001320511.1
NP_001307440.1
7
1167
NM_001320512.1
NP_001307441.1
6
1133
NM_001320513.1
NP_001307442.1
7
1545
NM_178177.4
NP_835471.1
5
984
XM_011512790.1
XP_011511092.1
4
559
XM_011512791.2
XP_011511093.1
4
858
XM_017006328.1
XP_016861817.1
8
1432
XM_017006329.1
XP_016861818.1
8
1429
XM_017006330.1
XP_016861819.1
7
1392
XM_017006331.1
XP_016861820.1
8
1505
XM_017006332.1
XP_016861821.1
8
5446
XM_017006333.1
XP_016861822.1
6
1295
XM_017006334.1
XP_016861823.1
6
1292
XM_017006335.1
XP_016861824.1
7
1144
XM_017006336.1
XP_016861825.1
6
1104
XM_017006337.1
XP_016861826.1
5
1007
XM_017006338.1
XP_016861827.1
4
559
XM_017006339.1
XP_016861828.1
6
1247
XM_017006340.1
XP_016861829.1
5
1216
XM_017006341.1
XP_016861830.1
7
1175
XM_017006342.1
XP_016861831.1
8
1281
XM_017006343.1
XP_016861832.1
7
1241
XM_017006344.1
XP_016861833.1
6
1141
GenBank mRNA
AF345564.1
-
4
683
AK123208.1
-
4
781
AK127477.1
-
2
2969
BC034374.1
-
5
932
5185621
BC036218.1
-
7
1512
5270916
BX649063.1
-
2
1814
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotidyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
NAD biosynthetic process
response to wounding
NAD metabolic process
'de novo' NAD biosynthetic process from aspartate
nicotinamide-nucleotide adenylyltransferase activity
nicotinate-nucleotide adenylyltransferase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline