Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006623.3 | NP_006614.2 | 12 | 1978 | ||
XM_011541226.1 | XP_011539528.1 | 14 | 2259 | |||
XM_011541227.2 | XP_011539529.1 | 14 | 1964 | |||
XM_011541228.1 | XP_011539530.1 | 14 | 1877 | |||
XM_011541231.2 | XP_011539533.1 | 14 | 2081 | |||
GenBank mRNA | AF171235.1 | - | 5 | 842 | ||
AF171236.1 | - | 9 | 1368 | |||
AF171237.1 | - | 12 | 1824 | |||
AK093306.1 | - | 11 | 3156 | |||
AK129762.1 | - | 4 | 1728 | |||
AK309662.1 | - | 14 | 2172 | |||
BC000303.2 | - | 12 | 1818 | 2820369 | ||
BC001349.2 | - | 12 | 1813 | 2961389 | ||
BC011262.1 | - | 12 | 1887 | 4156703 | ||
BC023235.1 | - | 12 | 1791 | |||
BC032110.2 | - | 6 | 1119 | |||
FJ224347.1 | - | 2443 |