Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178434.2 | NP_848521.1 | 1 | 251 | ||
GenBank mRNA | BC130365.1 | - | 1 | 251 | 40146396 | |
BC130367.1 | - | 1 | 251 | 40146398 |