Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206389.1 | NP_001193318.1 | 4, 5 | 373 | ||
NM_006119.4 | NP_006110.1 | 4, 5 | 466 | |||
NM_033163.3 | NP_149353.1 | 5, 6 | 553 | |||
NM_033164.3 | NP_149354.1 | 5, 6 | 520 | |||
NM_033165.3 | NP_149355.1 | 4, 5 | 433 | |||
GenBank mRNA | AB014615.1 | - | 5, 6 | 460 | ||
BC069106.1 | - | 5, 6 | 465 | 7216922 | ||
BC128235.1 | - | 4, 5 | 433 | 40114671 | ||
BC128236.1 | - | 4, 5 | 433 | 40114672 | ||
CN369230.1 | - | 4, 5 | 383 | |||
U36223.1 | - | 4, 5 | 373 | |||
U46211.1 | - | 4, 5 | 331 | |||
U46212.1 | - | 4, 5 | 364 | |||
U46213.1 | - | 5, 6 | 418 | |||
U56978.1 | - | 3, 4 | 336 |