Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP12-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181686.1 | NP_859014.1 | 1 | 470 | ||
GenBank mRNA | AB076363.1 | - | 1 | 450 | ||
AJ566388.1 | - | 1 | 470 |