Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021649.7 | NP_067681.1 | 2 | 930 | ||
GenBank mRNA | AB091054.1 | - | 1 | 491 | ||
AB097022.1 | - | 2 | 598 | |||
AB109098.1 | - | 2 | 574 | |||
AB446491.1 | - | 1 | 488 | |||
AY232653.1 | - | 1 | 488 | |||
AY304582.1 | - | 1 | 488 | |||
BC109265.2 | - | 2 | 830 | 40025951 | ||
BC109266.2 | - | 2 | 830 | 40025952 |
Gene Symbol: TMED7-TICAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164468.3 | NP_001157940.1 | 4 | 1382 | ||
NM_001164469.3 | NP_001157941.1 | 4 | 1500 | |||
GenBank mRNA | AK075218.1 | - | 4 | 1357 | ||
AY275836.1 | - | 3 | 702 | |||
AY304581.1 | - | 4 | 995 | |||
AY304583.1 | - | 4 | 1335 | |||
AY304584.1 | - | 4 | 1217 |