Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021250.3 | NP_067073.1 | 5 | 622 | ||
NM_181879.2 | NP_870994.1 | 5 | 622 | |||
NM_181985.3 | NP_871714.1 | 4 | 586 | |||
NM_181986.2 | NP_871715.1 | 4 | 586 | |||
GenBank mRNA | AF212842.1 | - | 2 | 336 | ||
AF324830.1 | - | 5 | 622 | |||
AF499916.1 | - | 5 | 565 | |||
AF499917.1 | - | 4 | 521 | |||
AF499918.1 | - | 5 | 581 | |||
AF499919.1 | - | 4 | 538 | |||
EU915611.1 | - | 5 | 562 | |||
EU915612.1 | - | 4 | 526 |