Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194249.2 | NP_919225.1 | 3 | 308 | ||
GenBank mRNA | AY321065.1 | - | 3 | 264 | ||
BC033496.1 | - | 3 | 308 | 5172595 |