Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198515.2 | NP_940917.1 | 2, 3 | 323 | ||
XM_006717820.3 | XP_006717883.1 | 2, 3 | 654 | |||
XM_011539771.2 | XP_011538073.1 | 2, 3 | 651 | |||
XM_017016206.1 | XP_016871695.1 | 2, 3 | 241 | |||
GenBank mRNA | AJ457798.1 | - | 1, 2 | 305 | ||
BC044830.1 | - | 2, 3 | 323 | 5166167 |